Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005560.4 | NP_005551.3 | 44 - 45 | 5915 | 71 | |
XM_006723796.3 | XP_006723859.1 | 44 - 45 | 5918 | 71 | ||
XM_006723798.3 | XP_006723861.1 | 44 - 45 | 5921 | 71 | ||
XM_011528818.2 | XP_011527120.1 | 43 - 44 | 5780 | 71 | ||
XM_011528819.2 | XP_011527121.1 | 42 - 43 | 5645 | 71 | ||
GenBank mRNA | AF443072.1 | - | 44 - 45 | 5915 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Probe spans exons |