Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021658.hg.1 |
TC06001796.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021658.hg.1 |
ENST00000340994.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021658.hg.1 |
ENST00000371117.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021658.hg.1 |
ENST00000393616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021658.hg.1 |
NM_138694.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021658.hg.1 |
NM_170724.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182773.hg.1 |
ENST00000340994.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182773.hg.1 |
ENST00000371117.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182773.hg.1 |
NM_138694.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182773.hg.1 |
NM_170724.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182773.hg.1 |
ENST00000340994.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182773.hg.1 |
ENST00000371117.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182773.hg.1 |
NM_138694.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182773.hg.1 |
NM_170724.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182773.hg.1 |
TC0600012032.hg.1 |
Clariom D TC |