Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4000492.hg.1 |
TC6_mann_hap4000036.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4000492.hg.1 |
NM_014046.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000259873.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000259873.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000259873.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000259873.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000259873.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000259873.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000259873.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000259873.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000259873.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000259873.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000472229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000472229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000472229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000472229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000472229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000472229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000472229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000472229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000472229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000472229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000472267.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000472267.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000472267.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000472267.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000472267.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000472267.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000472267.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000472267.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000472267.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000472267.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000472267.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000472267.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000492316.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000492316.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000492316.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000492316.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
ENST00000492316.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
NM_014046.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
NM_014046.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
NM_014046.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
NM_014046.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
NM_014046.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
NM_014046.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
NM_014046.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
NM_014046.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
NM_014046.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014097.hg.1 |
NM_014046.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3000501.hg.1 |
TC6_dbb_hap3000034.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3000501.hg.1 |
NM_014046.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6000498.hg.1 |
TC6_qbl_hap6000034.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6000498.hg.1 |
NM_014046.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000401.hg.1 |
TC6_mcf_hap5000028.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000401.hg.1 |
NM_014046.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002971.hg.1 |
TC06000343.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002971.hg.1 |
ENST00000259873.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002971.hg.1 |
ENST00000472229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002971.hg.1 |
ENST00000506373.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002971.hg.1 |
NM_014046.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000495.hg.1 |
TC6_ssto_hap7000034.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000495.hg.1 |
NM_014046.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000512.hg.1 |
TC6_cox_hap2000041.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000512.hg.1 |
NM_014046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000259873.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000259873.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000259873.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000259873.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000259873.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000259873.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000259873.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000259873.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000259873.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000259873.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000472229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000472229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000472229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000472229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000472229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000472229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000472229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000472229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000472229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000472229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000472267.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000472267.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000472267.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000472267.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000472267.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000472267.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000472267.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000472267.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000472267.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000472267.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000472267.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000472267.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000492316.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000492316.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000492316.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000492316.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
ENST00000492316.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
NM_014046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
NM_014046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
NM_014046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
NM_014046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
NM_014046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
NM_014046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
NM_014046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
NM_014046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
NM_014046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014097.hg.1 |
NM_014046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600197963.hg.1 |
ENST00000259873.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600197963.hg.1 |
ENST00000472229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600197963.hg.1 |
ENST00000472267.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600197963.hg.1 |
NM_014046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600197963.hg.1 |
ENST00000259873.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600197963.hg.1 |
ENST00000472229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600197963.hg.1 |
ENST00000472267.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600197963.hg.1 |
NM_014046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600197963.hg.1 |
TC0600014097.hg.1 |
Clariom D TC |