Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000429987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000429987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000429987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000429987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000429987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000429987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000429987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000429987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000429987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000429987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000429987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000430857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000430857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000430857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000430857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000430857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000430857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000430857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000430857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000430857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000430857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000474713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000474713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000474713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000474713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000474713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000474713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000474713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000474713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000474713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000474713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000474713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000474713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000474713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000477022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000477022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000477022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000477022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000477022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000477022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000477022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000477022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
ENST00000477022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009524.hg.1 |
TC07000625.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009524.hg.1 |
ENST00000262935.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009524.hg.1 |
ENST00000300176.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009524.hg.1 |
ENST00000474713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009524.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160198.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160198.hg.1 |
ENST00000429987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160198.hg.1 |
ENST00000430857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160198.hg.1 |
ENST00000474713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160198.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160198.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160198.hg.1 |
ENST00000429987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160198.hg.1 |
ENST00000430857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160198.hg.1 |
ENST00000474713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160198.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160198.hg.1 |
TC0700008552.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000300176.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000429987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000429987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000429987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000429987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000429987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000429987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000429987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000429987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000429987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000429987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000429987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000430857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000430857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000430857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000430857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000430857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000430857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000430857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000430857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000430857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000430857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000474713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000474713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000474713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000474713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000474713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000474713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000474713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000474713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000474713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000474713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000474713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000474713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000474713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000477022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000477022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000477022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000477022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000477022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000477022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000477022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000477022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
ENST00000477022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008552.hg.1 |
NM_006076.4 |
Refseq |