Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001184785.1 | NP_001171714.1 | 15 - 16 | 2547 | 117 | |
NM_001184786.1 | NP_001171715.1 | 14 - 15 | 2508 | 117 | ||
NM_001184788.1 | NP_001171717.1 | 14 - 15 | 2508 | 117 | ||
NM_001184789.1 | NP_001171718.1 | 14 - 15 | 2508 | 117 | ||
NM_001184790.1 | NP_001171719.1 | 13 - 14 | 2376 | 117 | ||
NM_001184791.1 | NP_001171720.1 | 13 - 14 | 2376 | 117 | ||
NM_001184792.1 | NP_001171721.1 | 15 - 16 | 2547 | 117 | ||
NM_001184793.1 | NP_001171722.1 | 15 - 16 | 2547 | 117 | ||
NM_001184794.1 | NP_001171723.1 | 14 - 15 | 2508 | 117 | ||
XM_005252528.4 | XP_005252585.1 | 15 - 16 | 2538 | 117 | ||
XM_005252530.4 | XP_005252587.1 | 15 - 16 | 2538 | 117 | ||
XM_005252531.4 | XP_005252588.1 | 15 - 16 | 2538 | 117 | ||
XM_005252532.4 | XP_005252589.1 | 14 - 15 | 2192 | 117 | ||
XM_005252534.4 | XP_005252591.1 | 15 - 16 | 2538 | 117 | ||
XM_005252535.4 | XP_005252592.1 | 14 - 15 | 2499 | 117 | ||
XM_017016430.1 | XP_016871919.1 | 15 - 16 | 2538 | 117 | ||
GenBank mRNA | AB073671.1 | - | 13 - 14 | 2046 | 117 | |
AF196185.1 | - | 15 - 16 | 2504 | 117 | ||
AF196186.1 | - | 15 - 16 | 2504 | 117 | ||
AF332592.1 | - | 14 - 15 | 2178 | 117 | ||
AF332593.1 | - | 14 - 15 | 2178 | 117 | ||
AF454057.1 | - | 13 - 14 | 1924 | 117 | ||
AF454059.1 | - | 10 - 11 | 1511 | 117 | ||
AK024668.1 | - | 2 - 3 | 199 | 117 | ||
AK025892.1 | - | 6 - 7 | 994 | 117 | ||
AK027735.1 | - | 13 - 14 | 1844 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |