Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X004778.hg.1 |
TC0X000371.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X004778.hg.1 |
ENST00000333026.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X004778.hg.1 |
NM_198512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X004781.hg.1 |
TC0X000371.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X004781.hg.1 |
ENST00000333026.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X004781.hg.1 |
NM_198512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X004780.hg.1 |
TC0X000371.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X004780.hg.1 |
ENST00000333026.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X004780.hg.1 |
NM_198512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007531.hg.1 |
ENST00000333026.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007531.hg.1 |
ENST00000333026.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007531.hg.1 |
ENST00000333026.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007531.hg.1 |
ENST00000333026.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007531.hg.1 |
ENST00000333026.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007531.hg.1 |
ENST00000333026.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007531.hg.1 |
ENST00000333026.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007531.hg.1 |
ENST00000333026.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007531.hg.1 |
ENST00000333026.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007531.hg.1 |
NM_198512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007531.hg.1 |
NM_198512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007531.hg.1 |
NM_198512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007531.hg.1 |
NM_198512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007531.hg.1 |
NM_198512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007531.hg.1 |
NM_198512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007531.hg.1 |
NM_198512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007531.hg.1 |
NM_198512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007531.hg.1 |
NM_198512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007531.hg.1 |
NM_198512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X004779.hg.1 |
TC0X000371.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X004779.hg.1 |
ENST00000333026.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X004779.hg.1 |
NM_198512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007531.hg.1 |
ENST00000333026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007531.hg.1 |
ENST00000333026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007531.hg.1 |
ENST00000333026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007531.hg.1 |
ENST00000333026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007531.hg.1 |
ENST00000333026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007531.hg.1 |
ENST00000333026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007531.hg.1 |
ENST00000333026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007531.hg.1 |
ENST00000333026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007531.hg.1 |
ENST00000333026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007531.hg.1 |
NM_198512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007531.hg.1 |
NM_198512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007531.hg.1 |
NM_198512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007531.hg.1 |
NM_198512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007531.hg.1 |
NM_198512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007531.hg.1 |
NM_198512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007531.hg.1 |
NM_198512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007531.hg.1 |
NM_198512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007531.hg.1 |
NM_198512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007531.hg.1 |
NM_198512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00151558.hg.1 |
ENST00000333026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00151558.hg.1 |
NM_198512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00151558.hg.1 |
ENST00000333026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00151558.hg.1 |
NM_198512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00151558.hg.1 |
TC0X00007531.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00151559.hg.1 |
ENST00000333026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00151559.hg.1 |
NM_198512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00151559.hg.1 |
ENST00000333026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00151559.hg.1 |
NM_198512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00151559.hg.1 |
TC0X00007531.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00151556.hg.1 |
ENST00000333026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00151556.hg.1 |
NM_198512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00151556.hg.1 |
ENST00000333026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00151556.hg.1 |
NM_198512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00151556.hg.1 |
TC0X00007531.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00151560.hg.1 |
ENST00000333026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00151560.hg.1 |
NM_198512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00151560.hg.1 |
ENST00000333026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00151560.hg.1 |
NM_198512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00151560.hg.1 |
TC0X00007531.hg.1 |
Clariom D TC |