Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012546.hg.1 |
ENST00000308924.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012546.hg.1 |
ENST00000450554.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012546.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012546.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012546.hg.1 |
TC19000906.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000308924.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000308924.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000308924.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000308924.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000308924.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000308924.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000308924.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000308924.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000308924.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000308924.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000308924.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000308924.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000308924.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000308924.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000587196.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000587196.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000587196.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000587196.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000587196.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000587196.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000587196.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000587196.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000587275.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000587275.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000587275.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000588850.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000588850.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000588850.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000588850.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000588850.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000588850.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000590551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000590551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000590551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000590551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000590551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000590551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000590551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000590551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000592867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000592867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000592867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000592874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000592874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000592874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000592874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000592874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000592874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000592874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
ENST00000592874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900170033.hg.1 |
ENST00000308924.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170033.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170033.hg.1 |
ENST00000587275.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170033.hg.1 |
ENST00000590551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170033.hg.1 |
ENST00000592874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170033.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900170033.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900170033.hg.1 |
TC1900008924.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900170033.hg.1 |
ENST00000308924.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170033.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170033.hg.1 |
ENST00000587275.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170033.hg.1 |
ENST00000590551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170033.hg.1 |
ENST00000592874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170033.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900170033.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000308924.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000308924.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000308924.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000308924.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000308924.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000308924.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000308924.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000308924.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000308924.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000308924.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000308924.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000308924.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000308924.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000308924.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000450554.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000587196.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000587196.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000587196.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000587196.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000587196.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000587196.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000587196.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000587196.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000587275.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000587275.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000587275.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000588850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000588850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000588850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000588850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000588850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000588850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000590551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000590551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000590551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000590551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000590551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000590551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000590551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000590551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000592867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000592867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000592867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000592874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000592874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000592874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000592874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000592874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000592874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000592874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
ENST00000592874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_001012478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008924.hg.1 |
NM_007279.2 |
Refseq |