Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001324417.1 | NP_001311346.1 | 26 - 27 | 2619 | 137 | |
NM_001324421.1 | NP_001311350.1 | 26 - 27 | 2622 | 137 | ||
NM_016351.5 | NP_057435.2 | 26 - 27 | 2622 | 137 | ||
NM_021721.4 | NP_068367.1 | 26 - 27 | 2622 | 137 | ||
NM_021722.5 | NP_068368.2 | 26 - 27 | 2622 | 137 | ||
XM_011516323.1 | XP_011514625.1 | 26 - 27 | 2514 | 137 | ||
XM_017012329.1 | XP_016867818.1 | 27 - 28 | 2565 | 137 | ||
XM_017012331.1 | XP_016867820.1 | 26 - 27 | 2595 | 137 | ||
XM_017012334.1 | XP_016867823.1 | 27 - 28 | 2565 | 137 | ||
XM_017012335.1 | XP_016867824.1 | 27 - 28 | 2565 | 137 | ||
XM_017012336.1 | XP_016867825.1 | 26 - 27 | 2514 | 137 | ||
XM_017012337.1 | XP_016867826.1 | 27 - 28 | 2565 | 137 | ||
XM_017012339.1 | XP_016867828.1 | 27 - 28 | 2565 | 137 | ||
XM_017012341.1 | XP_016867830.1 | 25 - 26 | 2242 | 137 | ||
GenBank mRNA | AF073291.2 | - | 26 - 27 | 2345 | 137 | |
AF155381.1 | - | 26 - 27 | 2345 | 137 | ||
AK291010.1 | - | 26 - 27 | 2469 | 137 | ||
BC125118.1 | - | 26 - 27 | 2376 | 40118904 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Homo sapiens Probe spans exons |