Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005517.hg.1 |
ENST00000244891.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005517.hg.1 |
ENST00000449290.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005517.hg.1 |
NM_032681.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005517.hg.1 |
TC11000442.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005519.hg.1 |
ENST00000244891.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005519.hg.1 |
ENST00000449290.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005519.hg.1 |
NM_032681.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005519.hg.1 |
TC11000442.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008482.hg.1 |
ENST00000447357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008482.hg.1 |
ENST00000447357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008482.hg.1 |
ENST00000603193.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008482.hg.1 |
ENST00000603193.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008482.hg.1 |
ENST00000603193.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008482.hg.1 |
ENST00000604755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008482.hg.1 |
ENST00000604755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011920.hg.1 |
ENST00000524374.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011920.hg.1 |
ENST00000524374.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011920.hg.1 |
ENST00000524374.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011920.hg.1 |
ENST00000524374.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011920.hg.1 |
ENST00000524374.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011920.hg.1 |
ENST00000524374.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005513.hg.1 |
ENST00000449290.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005513.hg.1 |
NM_032681.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005513.hg.1 |
TC11000442.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005516.hg.1 |
ENST00000244891.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005516.hg.1 |
ENST00000449290.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005516.hg.1 |
NM_032681.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005516.hg.1 |
TC11000442.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007619.hg.1 |
ENST00000244891.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007619.hg.1 |
ENST00000244891.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007619.hg.1 |
ENST00000244891.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007619.hg.1 |
ENST00000244891.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007619.hg.1 |
ENST00000244891.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007619.hg.1 |
ENST00000244891.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007619.hg.1 |
ENST00000449290.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007619.hg.1 |
ENST00000449290.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007619.hg.1 |
ENST00000449290.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007619.hg.1 |
ENST00000449290.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007619.hg.1 |
ENST00000449290.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007619.hg.1 |
ENST00000449290.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007619.hg.1 |
NM_032681.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007619.hg.1 |
NM_032681.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007619.hg.1 |
NM_032681.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007619.hg.1 |
NM_032681.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007619.hg.1 |
NM_032681.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007619.hg.1 |
NM_032681.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005512.hg.1 |
ENST00000449290.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005512.hg.1 |
NM_032681.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005512.hg.1 |
TC11000442.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008721.hg.1 |
ENST00000525732.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008721.hg.1 |
ENST00000525732.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008721.hg.1 |
ENST00000525732.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008721.hg.1 |
ENST00000525732.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008721.hg.1 |
ENST00000525732.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010822.hg.1 |
ENST00000526016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010822.hg.1 |
ENST00000526016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010822.hg.1 |
ENST00000526016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010822.hg.1 |
ENST00000526016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010822.hg.1 |
ENST00000526016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010822.hg.1 |
ENST00000530412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010822.hg.1 |
ENST00000530412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010822.hg.1 |
ENST00000530412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010822.hg.1 |
ENST00000530412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010822.hg.1 |
ENST00000530412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010822.hg.1 |
ENST00000530412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010822.hg.1 |
ENST00000530412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010822.hg.1 |
ENST00000530412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010822.hg.1 |
ENST00000530412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010822.hg.1 |
NR_038174.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010822.hg.1 |
NR_038174.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010822.hg.1 |
NR_038174.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010822.hg.1 |
NR_038174.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010822.hg.1 |
NR_038174.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010822.hg.1 |
NR_038174.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010822.hg.1 |
NR_038174.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005511.hg.1 |
ENST00000449290.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005511.hg.1 |
NM_032681.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005511.hg.1 |
TC11000442.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010784.hg.1 |
ENST00000534741.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010784.hg.1 |
ENST00000534741.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010784.hg.1 |
ENST00000534741.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010784.hg.1 |
ENST00000534741.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010784.hg.1 |
ENST00000534741.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007533.hg.1 |
ENST00000505076.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007533.hg.1 |
ENST00000505076.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007533.hg.1 |
ENST00000505076.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007533.hg.1 |
ENST00000505076.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007533.hg.1 |
ENST00000505076.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010799.hg.1 |
ENST00000526452.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010799.hg.1 |
ENST00000526452.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010799.hg.1 |
ENST00000526452.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010799.hg.1 |
ENST00000526452.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010799.hg.1 |
ENST00000526452.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010799.hg.1 |
ENST00000526452.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010799.hg.1 |
ENST00000526452.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010799.hg.1 |
ENST00000526452.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010799.hg.1 |
ENST00000526452.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153801.hg.1 |
ENST00000449290.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153801.hg.1 |
NM_032681.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153801.hg.1 |
ENST00000449290.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153801.hg.1 |
NM_032681.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153801.hg.1 |
TC1100007619.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100153807.hg.1 |
ENST00000244891.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153807.hg.1 |
ENST00000449290.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153807.hg.1 |
NM_032681.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153807.hg.1 |
ENST00000244891.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153807.hg.1 |
ENST00000449290.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153807.hg.1 |
NM_032681.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153807.hg.1 |
TC1100007619.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100007619.hg.1 |
ENST00000244891.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007619.hg.1 |
ENST00000244891.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007619.hg.1 |
ENST00000244891.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007619.hg.1 |
ENST00000244891.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007619.hg.1 |
ENST00000244891.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007619.hg.1 |
ENST00000244891.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007619.hg.1 |
ENST00000449290.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007619.hg.1 |
ENST00000449290.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007619.hg.1 |
ENST00000449290.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007619.hg.1 |
ENST00000449290.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007619.hg.1 |
ENST00000449290.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007619.hg.1 |
ENST00000449290.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007619.hg.1 |
NM_032681.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007619.hg.1 |
NM_032681.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007619.hg.1 |
NM_032681.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007619.hg.1 |
NM_032681.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007619.hg.1 |
NM_032681.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007619.hg.1 |
NM_032681.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010784.hg.1 |
ENST00000534741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010784.hg.1 |
ENST00000534741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010784.hg.1 |
ENST00000534741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010784.hg.1 |
ENST00000534741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010784.hg.1 |
ENST00000534741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153805.hg.1 |
ENST00000244891.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153805.hg.1 |
ENST00000449290.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153805.hg.1 |
NM_032681.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153805.hg.1 |
ENST00000244891.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153805.hg.1 |
ENST00000449290.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153805.hg.1 |
NM_032681.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153805.hg.1 |
TC1100007619.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100153800.hg.1 |
ENST00000449290.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153800.hg.1 |
NM_032681.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153800.hg.1 |
ENST00000449290.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153800.hg.1 |
NM_032681.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153800.hg.1 |
TC1100007619.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100153804.hg.1 |
ENST00000244891.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153804.hg.1 |
ENST00000449290.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153804.hg.1 |
NM_032681.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153804.hg.1 |
ENST00000244891.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153804.hg.1 |
ENST00000449290.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153804.hg.1 |
NM_032681.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153804.hg.1 |
TC1100007619.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100008721.hg.1 |
ENST00000525732.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008721.hg.1 |
ENST00000525732.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008721.hg.1 |
ENST00000525732.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008721.hg.1 |
ENST00000525732.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008721.hg.1 |
ENST00000525732.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153803.hg.1 |
ENST00000244891.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153803.hg.1 |
ENST00000244891.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153803.hg.1 |
TC1100007619.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100010799.hg.1 |
ENST00000526452.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010799.hg.1 |
ENST00000526452.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010799.hg.1 |
ENST00000526452.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010799.hg.1 |
ENST00000526452.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010799.hg.1 |
ENST00000526452.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010799.hg.1 |
ENST00000526452.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010799.hg.1 |
ENST00000526452.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010799.hg.1 |
ENST00000526452.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010799.hg.1 |
ENST00000526452.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010822.hg.1 |
ENST00000526016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010822.hg.1 |
ENST00000526016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010822.hg.1 |
ENST00000526016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010822.hg.1 |
ENST00000526016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010822.hg.1 |
ENST00000526016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010822.hg.1 |
ENST00000530412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010822.hg.1 |
ENST00000530412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010822.hg.1 |
ENST00000530412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010822.hg.1 |
ENST00000530412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010822.hg.1 |
ENST00000530412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010822.hg.1 |
ENST00000530412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010822.hg.1 |
ENST00000530412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010822.hg.1 |
ENST00000530412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010822.hg.1 |
ENST00000530412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010822.hg.1 |
NR_038174.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010822.hg.1 |
NR_038174.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010822.hg.1 |
NR_038174.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010822.hg.1 |
NR_038174.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010822.hg.1 |
NR_038174.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010822.hg.1 |
NR_038174.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010822.hg.1 |
NR_038174.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008482.hg.1 |
ENST00000447357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008482.hg.1 |
ENST00000447357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008482.hg.1 |
ENST00000603193.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008482.hg.1 |
ENST00000603193.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008482.hg.1 |
ENST00000603193.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008482.hg.1 |
ENST00000604755.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008482.hg.1 |
ENST00000604755.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007533.hg.1 |
ENST00000505076.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007533.hg.1 |
ENST00000505076.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007533.hg.1 |
ENST00000505076.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007533.hg.1 |
ENST00000505076.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007533.hg.1 |
ENST00000505076.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011920.hg.1 |
ENST00000524374.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011920.hg.1 |
ENST00000524374.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011920.hg.1 |
ENST00000524374.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011920.hg.1 |
ENST00000524374.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011920.hg.1 |
ENST00000524374.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011920.hg.1 |
ENST00000524374.1 |
Ensembl |