Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014610.hg.1 |
TC08001139.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014610.hg.1 |
ENST00000307599.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014610.hg.1 |
ENST00000518826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014610.hg.1 |
NM_152413.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010142.hg.1 |
ENST00000307599.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010142.hg.1 |
ENST00000307599.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010142.hg.1 |
ENST00000307599.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010142.hg.1 |
ENST00000307599.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010142.hg.1 |
ENST00000307599.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010142.hg.1 |
ENST00000307599.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010142.hg.1 |
ENST00000307599.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010142.hg.1 |
ENST00000307599.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010142.hg.1 |
ENST00000307599.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010142.hg.1 |
ENST00000307599.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010142.hg.1 |
ENST00000307599.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010142.hg.1 |
ENST00000307599.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010142.hg.1 |
ENST00000518826.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010142.hg.1 |
ENST00000518826.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010142.hg.1 |
ENST00000518826.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010142.hg.1 |
ENST00000518826.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010142.hg.1 |
ENST00000524298.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010142.hg.1 |
ENST00000524298.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010142.hg.1 |
ENST00000524298.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010142.hg.1 |
ENST00000524298.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010142.hg.1 |
ENST00000524298.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010142.hg.1 |
NM_152413.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010142.hg.1 |
NM_152413.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010142.hg.1 |
NM_152413.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010142.hg.1 |
NM_152413.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010142.hg.1 |
NM_152413.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010142.hg.1 |
NM_152413.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010142.hg.1 |
NM_152413.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010142.hg.1 |
NM_152413.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010142.hg.1 |
NM_152413.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010142.hg.1 |
NM_152413.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010142.hg.1 |
NM_152413.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010142.hg.1 |
NM_152413.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800168944.hg.1 |
ENST00000307599.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168944.hg.1 |
ENST00000518826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168944.hg.1 |
NM_152413.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800168944.hg.1 |
ENST00000307599.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168944.hg.1 |
ENST00000518826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168944.hg.1 |
NM_152413.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800168944.hg.1 |
TC0800010142.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800010142.hg.1 |
ENST00000307599.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010142.hg.1 |
ENST00000307599.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010142.hg.1 |
ENST00000307599.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010142.hg.1 |
ENST00000307599.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010142.hg.1 |
ENST00000307599.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010142.hg.1 |
ENST00000307599.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010142.hg.1 |
ENST00000307599.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010142.hg.1 |
ENST00000307599.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010142.hg.1 |
ENST00000307599.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010142.hg.1 |
ENST00000307599.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010142.hg.1 |
ENST00000307599.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010142.hg.1 |
ENST00000307599.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010142.hg.1 |
ENST00000518826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010142.hg.1 |
ENST00000518826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010142.hg.1 |
ENST00000518826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010142.hg.1 |
ENST00000518826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010142.hg.1 |
ENST00000524298.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010142.hg.1 |
ENST00000524298.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010142.hg.1 |
ENST00000524298.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010142.hg.1 |
ENST00000524298.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010142.hg.1 |
ENST00000524298.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010142.hg.1 |
NM_152413.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010142.hg.1 |
NM_152413.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010142.hg.1 |
NM_152413.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010142.hg.1 |
NM_152413.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010142.hg.1 |
NM_152413.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010142.hg.1 |
NM_152413.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010142.hg.1 |
NM_152413.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010142.hg.1 |
NM_152413.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010142.hg.1 |
NM_152413.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010142.hg.1 |
NM_152413.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010142.hg.1 |
NM_152413.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010142.hg.1 |
NM_152413.2 |
Refseq |