Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FREM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144966.5 | NP_659403.4 | 16 - 17 | 3458 | 68 | |
XM_005251382.3 | XP_005251439.1 | 15 - 16 | 3706 | 68 | ||
XM_017014316.1 | XP_016869805.1 | 15 - 16 | 3733 | 68 | ||
XM_017014317.1 | XP_016869806.1 | 16 - 17 | 3276 | 68 | ||
XM_017014318.1 | XP_016869807.1 | 16 - 17 | 4031 | 68 | ||
XM_017014319.1 | XP_016869808.1 | 17 - 18 | 3935 | 68 | ||
XM_017014320.1 | XP_016869809.1 | 16 - 17 | 3862 | 68 | ||
XM_017014321.1 | XP_016869810.1 | 15 - 16 | 3525 | 68 | ||
XM_017014322.1 | XP_016869811.1 | 15 - 16 | 3319 | 68 | ||
XM_017014323.1 | XP_016869812.1 | 15 - 16 | 2992 | 68 | ||
XM_017014324.1 | XP_016869813.1 | 15 - 16 | 3733 | 68 | ||
XM_017014325.1 | XP_016869814.1 | 15 - 16 | 3733 | 68 | ||
XM_017014326.1 | XP_016869815.1 | 14 - 15 | 2549 | 68 | ||
XM_017014327.1 | XP_016869816.1 | 11 - 12 | 1918 | 68 | ||
XM_017014328.1 | XP_016869817.1 | 15 - 16 | 3733 | 68 | ||
XM_017014329.1 | XP_016869818.1 | 15 - 16 | 3733 | 68 | ||
XM_017014330.1 | XP_016869819.1 | 15 - 16 | 3733 | 68 | ||
GenBank mRNA | AB160987.1 | - | 14 - 15 | 2642 | 68 | |
BX537574.1 | - | 16 - 17 | 3459 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |