Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000397076.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000397076.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000397076.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000397076.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000397076.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000397076.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000397076.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000397076.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000397076.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000397076.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000397076.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000397076.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000397076.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007250.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001620.hg.1 |
TC06000160.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001620.hg.1 |
ENST00000377905.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001620.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149196.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149196.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149196.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149196.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149196.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149196.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149196.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149196.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149196.hg.1 |
TC0600007250.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000377905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000397076.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000397076.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000397076.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000397076.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000397076.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000397076.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000397076.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000397076.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000397076.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000397076.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000397076.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000397076.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000397076.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
ENST00000439485.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
NM_001286121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
NM_005495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007250.hg.1 |
NM_005495.2 |
Refseq |