Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000246535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000246535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000246535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000246535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000246535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000246535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000246535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000246535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000246535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000246535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000585821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000585821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000585821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000585821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000587065.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000587065.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000587065.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000587065.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000587065.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000587065.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000587385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000587385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000587385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000587385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000587385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000587385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000587385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000587385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000587385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000587385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000589589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000589589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000589589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000589589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
ENST00000589589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
NM_032346.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
NM_032346.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
NM_032346.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
NM_032346.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
NM_032346.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
NM_032346.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
NM_032346.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
NM_032346.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
NM_032346.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011708.hg.1 |
NM_032346.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005789.hg.1 |
ENST00000246535.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005789.hg.1 |
NM_032346.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005789.hg.1 |
TC19000443.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900200344.hg.1 |
ENST00000246535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900200344.hg.1 |
ENST00000585821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900200344.hg.1 |
ENST00000587065.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900200344.hg.1 |
ENST00000587385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900200344.hg.1 |
ENST00000589589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900200344.hg.1 |
NM_032346.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900200344.hg.1 |
TC1900011708.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900200344.hg.1 |
ENST00000246535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900200344.hg.1 |
ENST00000585821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900200344.hg.1 |
ENST00000587065.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900200344.hg.1 |
ENST00000587385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900200344.hg.1 |
ENST00000589589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900200344.hg.1 |
NM_032346.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900200345.hg.1 |
ENST00000246535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900200345.hg.1 |
ENST00000587065.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900200345.hg.1 |
ENST00000587385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900200345.hg.1 |
ENST00000589589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900200345.hg.1 |
NM_032346.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900200345.hg.1 |
TC1900011708.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900200345.hg.1 |
ENST00000246535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900200345.hg.1 |
ENST00000587065.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900200345.hg.1 |
ENST00000587385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900200345.hg.1 |
ENST00000589589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900200345.hg.1 |
NM_032346.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000246535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000246535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000246535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000246535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000246535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000246535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000246535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000246535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000246535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000246535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000585821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000585821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000585821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000585821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000587065.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000587065.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000587065.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000587065.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000587065.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000587065.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000587385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000587385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000587385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000587385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000587385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000587385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000587385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000587385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000587385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000587385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000589589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000589589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000589589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000589589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
ENST00000589589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
NM_032346.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
NM_032346.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
NM_032346.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
NM_032346.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
NM_032346.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
NM_032346.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
NM_032346.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
NM_032346.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
NM_032346.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011708.hg.1 |
NM_032346.1 |
Refseq |