Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLFN14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129820.1 | NP_001123292.1 | 2 - 3 | 1218 | 118 | |
XM_017024576.1 | XP_016880065.1 | 4 - 5 | 1387 | 118 | ||
XM_017024577.1 | XP_016880066.1 | 4 - 5 | 1409 | 118 | ||
XM_017024578.1 | XP_016880067.1 | 3 - 4 | 1276 | 118 | ||
XM_017024579.1 | XP_016880068.1 | 3 - 4 | 1309 | 118 | ||
GenBank mRNA | BC140847.1 | - | 2 - 3 | 1215 | 9021715 | 118 |
BC157877.1 | - | 2 - 3 | 1219 | 9057094 | 118 | |
BC157879.1 | - | 2 - 3 | 1226 | 9057095 | 118 | |
BU658532.1 | - | 2 - 3 | 359 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |