Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
ENST00000312753.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
ENST00000312753.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
ENST00000312753.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
ENST00000312753.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
ENST00000312753.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
ENST00000312753.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
ENST00000312753.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
ENST00000312753.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
ENST00000312753.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
ENST00000361928.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
ENST00000361928.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
ENST00000361928.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
ENST00000361928.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
ENST00000361928.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
ENST00000361928.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
ENST00000581100.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
ENST00000581100.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
ENST00000581100.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
ENST00000581100.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
ENST00000581100.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
ENST00000581100.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
ENST00000604273.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
ENST00000604273.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
ENST00000604273.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
ENST00000604273.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
ENST00000604273.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
ENST00000604273.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
ENST00000619161.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
ENST00000619161.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
ENST00000619161.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
ENST00000619161.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
ENST00000619161.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
NR_002328.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
NR_002328.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
NR_002328.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
NR_002328.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
NR_002328.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
NR_002328.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
NR_002328.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
NR_002328.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
NR_002328.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
NR_002328.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
NR_104033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
NR_104033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
NR_104033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
NR_104033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
NR_104033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
NR_104033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
NR_104034.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
NR_104034.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
NR_104034.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009749.hg.1 |
NR_104034.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100170139.hg.1 |
ENST00000619161.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170139.hg.1 |
NR_104034.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100170139.hg.1 |
ENST00000619161.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170139.hg.1 |
NR_104034.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100170139.hg.1 |
TC0100009749.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100170133.hg.1 |
ENST00000312753.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170133.hg.1 |
ENST00000604273.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170133.hg.1 |
ENST00000619161.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170133.hg.1 |
NR_002328.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100170133.hg.1 |
NR_104034.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100170133.hg.1 |
ENST00000312753.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170133.hg.1 |
ENST00000604273.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170133.hg.1 |
ENST00000619161.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170133.hg.1 |
NR_002328.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100170133.hg.1 |
NR_104034.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100170133.hg.1 |
TC0100009749.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
ENST00000312753.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
ENST00000312753.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
ENST00000312753.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
ENST00000312753.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
ENST00000312753.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
ENST00000312753.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
ENST00000312753.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
ENST00000312753.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
ENST00000312753.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
ENST00000361928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
ENST00000361928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
ENST00000361928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
ENST00000361928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
ENST00000361928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
ENST00000361928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
ENST00000581100.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
ENST00000581100.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
ENST00000581100.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
ENST00000581100.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
ENST00000581100.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
ENST00000581100.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
ENST00000604273.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
ENST00000604273.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
ENST00000604273.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
ENST00000604273.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
ENST00000604273.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
ENST00000604273.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
ENST00000619161.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
ENST00000619161.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
ENST00000619161.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
ENST00000619161.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
ENST00000619161.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
NR_002328.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
NR_002328.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
NR_002328.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
NR_002328.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
NR_002328.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
NR_002328.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
NR_002328.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
NR_002328.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
NR_002328.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
NR_002328.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
NR_104033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
NR_104033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
NR_104033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
NR_104033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
NR_104033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
NR_104033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
NR_104034.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
NR_104034.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
NR_104034.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009749.hg.1 |
NR_104034.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100170134.hg.1 |
ENST00000312753.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170134.hg.1 |
ENST00000361928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170134.hg.1 |
ENST00000581100.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170134.hg.1 |
ENST00000604273.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170134.hg.1 |
ENST00000619161.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170134.hg.1 |
NR_002328.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100170134.hg.1 |
NR_104033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100170134.hg.1 |
NR_104034.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100170134.hg.1 |
ENST00000312753.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170134.hg.1 |
ENST00000361928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170134.hg.1 |
ENST00000581100.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170134.hg.1 |
ENST00000604273.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170134.hg.1 |
ENST00000619161.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170134.hg.1 |
NR_002328.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100170134.hg.1 |
NR_104033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100170134.hg.1 |
NR_104034.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100170134.hg.1 |
TC0100009749.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100170137.hg.1 |
ENST00000312753.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170137.hg.1 |
ENST00000361928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170137.hg.1 |
ENST00000581100.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170137.hg.1 |
ENST00000604273.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170137.hg.1 |
ENST00000619161.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170137.hg.1 |
NR_002328.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100170137.hg.1 |
NR_104033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100170137.hg.1 |
NR_104034.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100170137.hg.1 |
ENST00000312753.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170137.hg.1 |
ENST00000361928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170137.hg.1 |
ENST00000581100.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170137.hg.1 |
ENST00000604273.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170137.hg.1 |
ENST00000619161.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170137.hg.1 |
NR_002328.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100170137.hg.1 |
NR_104033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100170137.hg.1 |
NR_104034.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100170137.hg.1 |
TC0100009749.hg.1 |
Clariom D TC |