Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012540.hg.1 |
TC03000704.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012540.hg.1 |
ENST00000265379.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012540.hg.1 |
ENST00000432398.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012540.hg.1 |
NM_153264.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012540.hg.1 |
NR_022012.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012545.hg.1 |
TC03000704.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012545.hg.1 |
ENST00000265379.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012545.hg.1 |
ENST00000432398.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012545.hg.1 |
NM_153264.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012545.hg.1 |
NR_022012.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300162735.hg.1 |
ENST00000265379.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300162735.hg.1 |
ENST00000312481.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300162735.hg.1 |
ENST00000512836.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300162735.hg.1 |
NM_001278298.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300162735.hg.1 |
NM_153264.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300162735.hg.1 |
NR_022012.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300162735.hg.1 |
ENST00000265379.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300162735.hg.1 |
ENST00000312481.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300162735.hg.1 |
ENST00000512836.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300162735.hg.1 |
NM_001278298.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300162735.hg.1 |
NM_153264.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300162735.hg.1 |
NR_022012.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300162735.hg.1 |
TC0300008818.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300162731.hg.1 |
ENST00000265379.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300162731.hg.1 |
ENST00000312481.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300162731.hg.1 |
NM_001278298.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300162731.hg.1 |
NM_153264.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300162731.hg.1 |
NR_022012.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300162731.hg.1 |
ENST00000265379.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300162731.hg.1 |
ENST00000312481.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300162731.hg.1 |
NM_001278298.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300162731.hg.1 |
NM_153264.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300162731.hg.1 |
NR_022012.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300162731.hg.1 |
TC0300008818.hg.1 |
Clariom D TC |