Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DZIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014648.3 | NP_055463.1 | 28 - 29 | 3572 | 155 | |
XM_005247914.3 | XP_005247971.1 | 28 - 29 | 3371 | 155 | ||
XM_005247915.3 | XP_005247972.1 | 28 - 29 | 3341 | 155 | ||
XM_005247916.3 | XP_005247973.1 | 29 - 30 | 3513 | 155 | ||
XM_005247917.3 | XP_005247974.1 | 28 - 29 | 3302 | 155 | ||
XM_005247918.3 | XP_005247975.1 | 28 - 29 | 3789 | 155 | ||
XM_005247919.3 | XP_005247976.1 | 27 - 28 | 2923 | 155 | ||
XM_017007540.1 | XP_016863029.1 | 28 - 29 | 3418 | 155 | ||
GenBank mRNA | AB014575.1 | - | 28 - 29 | 3279 | 155 | |
AF279370.1 | - | 13 - 14 | 1231 | 155 | ||
AK023138.1 | - | 28 - 29 | 3340 | 155 | ||
AK294967.1 | - | 17 - 18 | 1635 | 155 | ||
AK303942.1 | - | 15 - 16 | 1725 | 155 | ||
AY495710.1 | - | 3 - 4 | 353 | 155 | ||
BC063882.1 | - | 28 - 29 | 3325 | 30387514 | 155 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |