Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014296.2 | NP_055111.1 | 16 - 17 | 2137 | 88 | |
XM_011533555.2 | XP_011531857.1 | 17 - 18 | 2036 | 88 | ||
XM_011533556.2 | XP_011531858.1 | 18 - 19 | 2268 | 88 | ||
XM_017006043.1 | XP_016861532.1 | 17 - 18 | 2220 | 88 | ||
GenBank mRNA | AB028639.1 | - | 16 - 17 | 2118 | 88 | |
AK314253.1 | - | 16 - 17 | 2099 | 88 | ||
BC056202.1 | - | 16 - 17 | 2115 | 5274889 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Homo sapiens Probe spans exons |