Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000457328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000457328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000470082.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000470082.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
ENST00000470082.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015274.hg.1 |
TC09001193.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015274.hg.1 |
ENST00000265381.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015274.hg.1 |
ENST00000470082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015274.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900170819.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900170819.hg.1 |
ENST00000470082.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900170819.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900170819.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900170819.hg.1 |
ENST00000470082.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900170819.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900170819.hg.1 |
TC0900010350.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000265381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000457328.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000457328.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000470082.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000470082.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
ENST00000470082.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010350.hg.1 |
NM_001163.3 |
Refseq |