Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000464133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000464133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000464133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000464133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000464133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000474172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000474172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000474172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000477049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000477049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000477049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000477049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000477049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000477049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000477049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
ENST00000477049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021316.hg.1 |
TC09001673.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021316.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021316.hg.1 |
ENST00000372169.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021316.hg.1 |
ENST00000393220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021316.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900180624.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180624.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180624.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900180624.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180624.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180624.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900180624.hg.1 |
TC0900011803.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000224140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000436441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000464133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000464133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000464133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000464133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000464133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000474172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000474172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000474172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000477049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000477049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000477049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000477049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000477049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000477049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000477049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
ENST00000477049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011803.hg.1 |
NM_015046.6 |
Refseq |