Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024657.hg.1 |
ENST00000251074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024657.hg.1 |
ENST00000543021.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024657.hg.1 |
ENST00000552283.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024657.hg.1 |
NM_024057.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024657.hg.1 |
TC12001887.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000251074.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000251074.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000251074.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000251074.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000251074.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000251074.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000251074.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000251074.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000251074.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000251074.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000251074.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000251074.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000251074.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000251074.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000543021.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000543021.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000543021.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000543021.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000543021.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000543021.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000543021.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000543021.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000543021.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000546385.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000546385.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000546385.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000546385.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000546385.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000546385.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000546385.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000547269.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000547269.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000547269.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000547269.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000547269.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000547269.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000547269.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000547269.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000548994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000548994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000548994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000548994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000548994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000550459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000550459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000550459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000551200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000551200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000551200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000551200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000551200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000551200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000551200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000551200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000551200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000551200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000551744.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000551744.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000551744.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000551744.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000551744.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000551744.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000551744.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000551744.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000551744.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000551744.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000551744.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
NM_024057.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
NM_024057.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
NM_024057.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
NM_024057.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
NM_024057.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
NM_024057.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
NM_024057.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
NM_024057.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
NM_024057.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
NM_024057.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
NM_024057.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
NM_024057.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
NM_024057.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011720.hg.1 |
NM_024057.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200190811.hg.1 |
ENST00000251074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190811.hg.1 |
ENST00000543021.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190811.hg.1 |
ENST00000551200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190811.hg.1 |
ENST00000551744.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190811.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190811.hg.1 |
NM_024057.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200190811.hg.1 |
ENST00000251074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190811.hg.1 |
ENST00000543021.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190811.hg.1 |
ENST00000551200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190811.hg.1 |
ENST00000551744.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190811.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190811.hg.1 |
NM_024057.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200190811.hg.1 |
TC1200011720.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000251074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000251074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000251074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000251074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000251074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000251074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000251074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000251074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000251074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000251074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000251074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000251074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000251074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000251074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000543021.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000543021.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000543021.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000543021.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000543021.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000543021.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000543021.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000543021.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000543021.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000546385.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000546385.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000546385.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000546385.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000546385.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000546385.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000546385.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000547269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000547269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000547269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000547269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000547269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000547269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000547269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000547269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000548994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000548994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000548994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000548994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000548994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000550459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000550459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000550459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000551200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000551200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000551200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000551200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000551200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000551200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000551200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000551200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000551200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000551200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000551744.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000551744.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000551744.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000551744.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000551744.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000551744.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000551744.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000551744.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000551744.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000551744.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000551744.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
ENST00000552283.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
NM_024057.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
NM_024057.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
NM_024057.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
NM_024057.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
NM_024057.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
NM_024057.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
NM_024057.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
NM_024057.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
NM_024057.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
NM_024057.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
NM_024057.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
NM_024057.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
NM_024057.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011720.hg.1 |
NM_024057.3 |
Refseq |