Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267059.1 | NP_001253988.1 | 3 - 4 | 472 | 90 | |
NM_001267060.1 | NP_001253989.1 | 2 - 3 | 433 | 90 | ||
NM_001267061.1 | NP_001253990.1 | 3 - 4 | 343 | 90 | ||
NM_014748.3 | NP_055563.1 | 3 - 4 | 508 | 90 | ||
NR_049782.1 | 3 - 4 | 633 | 90 | |||
GenBank mRNA | AK129689.1 | - | 3 - 4 | 392 | 90 | |
AK222543.1 | - | 3 - 4 | 391 | 90 | ||
AK293464.1 | - | 3 - 4 | 324 | 90 | ||
AK297054.1 | - | 3 - 4 | 517 | 90 | ||
AK298620.1 | - | 3 - 4 | 343 | 90 | ||
AK298869.1 | - | 2 - 3 | 294 | 90 | ||
AK300144.1 | - | 3 - 4 | 355 | 90 | ||
BC002524.1 | - | 3 - 4 | 369 | 3140610 | 90 | |
BC002610.1 | - | 3 - 4 | 358 | 3143104 | 90 | |
BC014620.2 | - | 3 - 4 | 369 | 3010648 | 90 | |
BC050590.1 | - | 3 - 4 | 363 | 6044270 | 90 | |
BT007167.1 | - | 3 - 4 | 260 | 90 | ||
CR457081.1 | - | 3 - 4 | 260 | 90 | ||
D31764.1 | - | 3 - 4 | 482 | 90 | ||
DB177194.1 | - | 3 - 4 | 498 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |