Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002592.hg.1 |
ENST00000382771.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002592.hg.1 |
NM_002639.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002592.hg.1 |
TC18000226.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000382771.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000382771.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000382771.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000382771.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000382771.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000382771.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000382771.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000382771.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000382771.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000382771.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000382771.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000382771.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000424602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000424602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000424602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000424602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000464346.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000464346.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000464346.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000464346.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000465652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000465652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000465652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000465652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000489441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000489441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000489441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000489441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000489441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000489441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000489441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000588986.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000588986.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000588986.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
ENST00000588986.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
NM_002639.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
NM_002639.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
NM_002639.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
NM_002639.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
NM_002639.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
NM_002639.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
NM_002639.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
NM_002639.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
NM_002639.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
NM_002639.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007543.hg.1 |
NM_002639.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000382771.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000382771.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000382771.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000382771.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000382771.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000382771.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000382771.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000382771.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000382771.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000382771.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000382771.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000382771.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000424602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000424602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000424602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000424602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000464346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000464346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000464346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000464346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000465652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000465652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000465652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000465652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000489441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000489441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000489441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000489441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000489441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000489441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000489441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000588986.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000588986.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000588986.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
ENST00000588986.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
NM_002639.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
NM_002639.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
NM_002639.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
NM_002639.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
NM_002639.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
NM_002639.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
NM_002639.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
NM_002639.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
NM_002639.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
NM_002639.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007543.hg.1 |
NM_002639.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800151747.hg.1 |
ENST00000382771.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151747.hg.1 |
ENST00000424602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151747.hg.1 |
ENST00000489441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151747.hg.1 |
ENST00000588986.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151747.hg.1 |
NM_002639.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800151747.hg.1 |
TC1800007543.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800151747.hg.1 |
ENST00000382771.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151747.hg.1 |
ENST00000424602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151747.hg.1 |
ENST00000489441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151747.hg.1 |
ENST00000588986.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151747.hg.1 |
NM_002639.4 |
Refseq |