Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000472375.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000472375.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000472375.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000472375.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000472375.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000476640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000476640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000476640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000476640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047490.hg.1 |
TC01003050.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047490.hg.1 |
ENST00000369406.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047490.hg.1 |
ENST00000476640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047490.hg.1 |
ENST00000544913.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047490.hg.1 |
NM_001166107.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047490.hg.1 |
NM_005518.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000472375.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000472375.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000472375.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000472375.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000472375.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000476640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000476640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000476640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000476640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215540.hg.1 |
ENST00000369406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215540.hg.1 |
ENST00000472375.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215540.hg.1 |
ENST00000476640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215540.hg.1 |
ENST00000544913.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215540.hg.1 |
NM_001166107.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215540.hg.1 |
NM_005518.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215540.hg.1 |
ENST00000369406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215540.hg.1 |
ENST00000472375.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215540.hg.1 |
ENST00000476640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215540.hg.1 |
ENST00000544913.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215540.hg.1 |
NM_001166107.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215540.hg.1 |
NM_005518.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215540.hg.1 |
TC0100015476.hg.1 |
Clariom D TC |