Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029371.hg.1 |
TC02001835.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029371.hg.1 |
ENST00000331040.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029371.hg.1 |
ENST00000401669.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029371.hg.1 |
ENST00000402717.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029371.hg.1 |
ENST00000404971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029371.hg.1 |
ENST00000405472.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029371.hg.1 |
ENST00000406316.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029371.hg.1 |
ENST00000406859.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029371.hg.1 |
ENST00000536085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029371.hg.1 |
NM_001135659.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029371.hg.1 |
NM_004801.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200191660.hg.1 |
ENST00000331040.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191660.hg.1 |
ENST00000401669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191660.hg.1 |
ENST00000402717.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191660.hg.1 |
ENST00000404971.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191660.hg.1 |
ENST00000405472.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191660.hg.1 |
ENST00000406316.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191660.hg.1 |
ENST00000406859.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191660.hg.1 |
ENST00000625672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191660.hg.1 |
ENST00000625891.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191660.hg.1 |
ENST00000626249.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191660.hg.1 |
ENST00000628515.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191660.hg.1 |
ENST00000628761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191660.hg.1 |
ENST00000630543.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191660.hg.1 |
NM_001135659.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200191660.hg.1 |
NM_004801.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200191660.hg.1 |
ENST00000331040.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191660.hg.1 |
ENST00000401669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191660.hg.1 |
ENST00000402717.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191660.hg.1 |
ENST00000404971.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191660.hg.1 |
ENST00000405472.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191660.hg.1 |
ENST00000406316.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191660.hg.1 |
ENST00000406859.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191660.hg.1 |
ENST00000625672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191660.hg.1 |
ENST00000625891.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191660.hg.1 |
ENST00000626249.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191660.hg.1 |
ENST00000628515.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191660.hg.1 |
ENST00000628761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191660.hg.1 |
ENST00000630543.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191660.hg.1 |
NM_001135659.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200191660.hg.1 |
NM_004801.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200191660.hg.1 |
TC0200012559.hg.1 |
Clariom D TC |