Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000302127.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000302127.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000302127.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000302127.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000302127.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000302127.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000302127.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000302127.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000302127.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000302127.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000302127.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000302127.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000315987.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000315987.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000315987.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000315987.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000315987.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000315987.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000315987.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000315987.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000315987.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000315987.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000315987.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000315987.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000315987.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000369697.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000369697.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000369697.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000369697.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000369697.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000369697.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000369697.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000369697.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000369697.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000369697.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000369697.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000369697.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000369697.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
ENST00000369697.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
NM_004980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
NM_004980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
NM_004980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
NM_004980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
NM_004980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
NM_004980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
NM_004980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
NM_004980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
NM_004980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
NM_004980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
NM_004980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
NM_004980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
NM_004980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
NM_172198.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
NM_172198.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
NM_172198.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
NM_172198.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
NM_172198.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
NM_172198.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
NM_172198.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
NM_172198.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
NM_172198.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
NM_172198.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
NM_172198.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015288.hg.1 |
NM_172198.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046437.hg.1 |
TC01002992.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046437.hg.1 |
ENST00000302127.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046437.hg.1 |
ENST00000315987.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046437.hg.1 |
ENST00000369697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046437.hg.1 |
NM_004980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046437.hg.1 |
NM_172198.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214009.hg.1 |
ENST00000302127.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214009.hg.1 |
ENST00000315987.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214009.hg.1 |
ENST00000369697.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214009.hg.1 |
NM_004980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214009.hg.1 |
NM_172198.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214009.hg.1 |
ENST00000302127.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214009.hg.1 |
ENST00000315987.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214009.hg.1 |
ENST00000369697.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214009.hg.1 |
NM_004980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214009.hg.1 |
NM_172198.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214009.hg.1 |
TC0100015288.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000302127.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000302127.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000302127.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000302127.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000302127.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000302127.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000302127.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000302127.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000302127.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000302127.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000302127.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000302127.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000315987.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000315987.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000315987.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000315987.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000315987.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000315987.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000315987.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000315987.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000315987.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000315987.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000315987.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000315987.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000315987.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000369697.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000369697.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000369697.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000369697.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000369697.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000369697.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000369697.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000369697.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000369697.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000369697.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000369697.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000369697.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000369697.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
ENST00000369697.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
NM_004980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
NM_004980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
NM_004980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
NM_004980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
NM_004980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
NM_004980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
NM_004980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
NM_004980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
NM_004980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
NM_004980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
NM_004980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
NM_004980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
NM_004980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
NM_172198.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
NM_172198.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
NM_172198.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
NM_172198.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
NM_172198.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
NM_172198.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
NM_172198.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
NM_172198.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
NM_172198.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
NM_172198.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
NM_172198.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015288.hg.1 |
NM_172198.2 |
Refseq |