Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047010.hg.1 |
TC01003014.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047010.hg.1 |
ENST00000369535.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047010.hg.1 |
NM_002524.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015352.hg.1 |
ENST00000369535.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015352.hg.1 |
ENST00000369535.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015352.hg.1 |
ENST00000369535.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015352.hg.1 |
ENST00000369535.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015352.hg.1 |
ENST00000369535.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015352.hg.1 |
ENST00000369535.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015352.hg.1 |
ENST00000369535.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015352.hg.1 |
ENST00000369535.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015352.hg.1 |
ENST00000369535.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015352.hg.1 |
NM_002524.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015352.hg.1 |
NM_002524.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015352.hg.1 |
NM_002524.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015352.hg.1 |
NM_002524.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015352.hg.1 |
NM_002524.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015352.hg.1 |
NM_002524.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015352.hg.1 |
NM_002524.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015352.hg.1 |
NM_002524.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015352.hg.1 |
NM_002524.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214710.hg.1 |
ENST00000369535.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214710.hg.1 |
NM_002524.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214710.hg.1 |
ENST00000369535.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214710.hg.1 |
NM_002524.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214710.hg.1 |
TC0100015352.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100015352.hg.1 |
ENST00000369535.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015352.hg.1 |
ENST00000369535.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015352.hg.1 |
ENST00000369535.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015352.hg.1 |
ENST00000369535.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015352.hg.1 |
ENST00000369535.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015352.hg.1 |
ENST00000369535.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015352.hg.1 |
ENST00000369535.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015352.hg.1 |
ENST00000369535.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015352.hg.1 |
ENST00000369535.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015352.hg.1 |
NM_002524.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015352.hg.1 |
NM_002524.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015352.hg.1 |
NM_002524.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015352.hg.1 |
NM_002524.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015352.hg.1 |
NM_002524.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015352.hg.1 |
NM_002524.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015352.hg.1 |
NM_002524.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015352.hg.1 |
NM_002524.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015352.hg.1 |
NM_002524.4 |
Refseq |