Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005309.hg.1 |
ENST00000262519.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005309.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005309.hg.1 |
TC16000364.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
ENST00000452917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
ENST00000452917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
ENST00000452917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
ENST00000452917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
ENST00000452917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
ENST00000452917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005311.hg.1 |
ENST00000262519.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005311.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005311.hg.1 |
TC16000364.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600154873.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600154873.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600154873.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600154873.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600154873.hg.1 |
TC1600007504.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
ENST00000262519.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
ENST00000452917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
ENST00000452917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
ENST00000452917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
ENST00000452917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
ENST00000452917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
ENST00000452917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007504.hg.1 |
NM_014712.2 |
Refseq |