Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017945.hg.1 |
TC06001546.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017945.hg.1 |
ENST00000375654.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017945.hg.1 |
ENST00000417199.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017945.hg.1 |
NM_005527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017947.hg.1 |
TC06001546.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017947.hg.1 |
ENST00000375653.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017947.hg.1 |
ENST00000375654.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017947.hg.1 |
ENST00000417199.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017947.hg.1 |
ENST00000424494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017947.hg.1 |
NM_005527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017946.hg.1 |
TC06001546.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017946.hg.1 |
ENST00000375653.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017946.hg.1 |
ENST00000375654.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017946.hg.1 |
ENST00000417199.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017946.hg.1 |
NM_005527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017943.hg.1 |
TC06001546.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017943.hg.1 |
ENST00000375653.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017943.hg.1 |
ENST00000375654.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017943.hg.1 |
ENST00000417199.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017943.hg.1 |
ENST00000424494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017943.hg.1 |
NM_005527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011459.hg.1 |
ENST00000375654.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011459.hg.1 |
ENST00000375654.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011459.hg.1 |
ENST00000375654.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011459.hg.1 |
ENST00000375654.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011459.hg.1 |
ENST00000375654.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011459.hg.1 |
NM_005527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011459.hg.1 |
NM_005527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011459.hg.1 |
NM_005527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011459.hg.1 |
NM_005527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011459.hg.1 |
NM_005527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600177718.hg.1 |
ENST00000375654.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177718.hg.1 |
NM_005527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600177718.hg.1 |
ENST00000375654.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177718.hg.1 |
NM_005527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600177718.hg.1 |
TC0600011459.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600011459.hg.1 |
ENST00000375654.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011459.hg.1 |
ENST00000375654.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011459.hg.1 |
ENST00000375654.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011459.hg.1 |
ENST00000375654.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011459.hg.1 |
ENST00000375654.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011459.hg.1 |
NM_005527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011459.hg.1 |
NM_005527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011459.hg.1 |
NM_005527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011459.hg.1 |
NM_005527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011459.hg.1 |
NM_005527.3 |
Refseq |