Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006671.hg.1 |
ENST00000291672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006671.hg.1 |
ENST00000330205.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006671.hg.1 |
NM_001142854.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006671.hg.1 |
NM_032261.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006671.hg.1 |
TC21000553.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
ENST00000291672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
ENST00000291672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
ENST00000291672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
ENST00000291672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
ENST00000291672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
ENST00000291672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
ENST00000291672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
ENST00000291672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
ENST00000291672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
ENST00000291672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
ENST00000291672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
ENST00000291672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
ENST00000330205.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
ENST00000330205.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
ENST00000330205.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
ENST00000330205.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
ENST00000330205.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
ENST00000330205.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
ENST00000330205.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
ENST00000330205.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
ENST00000330205.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
ENST00000330205.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
NM_001142854.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
NM_001142854.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
NM_001142854.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
NM_001142854.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
NM_001142854.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
NM_001142854.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
NM_001142854.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
NM_001142854.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
NM_001142854.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
NM_001142854.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
NM_001142854.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
NM_001142854.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
NM_032261.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
NM_032261.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
NM_032261.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
NM_032261.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
NM_032261.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
NM_032261.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
NM_032261.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
NM_032261.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
NM_032261.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008461.hg.1 |
NM_032261.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
ENST00000291672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
ENST00000291672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
ENST00000291672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
ENST00000291672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
ENST00000291672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
ENST00000291672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
ENST00000291672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
ENST00000291672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
ENST00000291672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
ENST00000291672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
ENST00000291672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
ENST00000291672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
ENST00000330205.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
ENST00000330205.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
ENST00000330205.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
ENST00000330205.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
ENST00000330205.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
ENST00000330205.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
ENST00000330205.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
ENST00000330205.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
ENST00000330205.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
ENST00000330205.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
NM_001142854.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
NM_001142854.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
NM_001142854.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
NM_001142854.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
NM_001142854.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
NM_001142854.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
NM_001142854.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
NM_001142854.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
NM_001142854.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
NM_001142854.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
NM_001142854.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
NM_001142854.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
NM_032261.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
NM_032261.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
NM_032261.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
NM_032261.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
NM_032261.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
NM_032261.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
NM_032261.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
NM_032261.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
NM_032261.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008461.hg.1 |
NM_032261.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100157334.hg.1 |
TC2100008461.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100157334.hg.1 |
ENST00000291672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157334.hg.1 |
ENST00000330205.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157334.hg.1 |
NM_001142854.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100157334.hg.1 |
NM_032261.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100157334.hg.1 |
ENST00000291672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157334.hg.1 |
ENST00000330205.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157334.hg.1 |
NM_001142854.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100157334.hg.1 |
NM_032261.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100157335.hg.1 |
TC2100008461.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100157335.hg.1 |
ENST00000291672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157335.hg.1 |
ENST00000330205.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157335.hg.1 |
NM_001142854.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100157335.hg.1 |
NM_032261.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100157335.hg.1 |
ENST00000291672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157335.hg.1 |
ENST00000330205.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157335.hg.1 |
NM_001142854.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100157335.hg.1 |
NM_032261.4 |
Refseq |