Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000571044.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000571044.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000571044.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000571044.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000571044.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000571044.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000571044.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000571044.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000571605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000571605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000571605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000571605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000571605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000571605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000571605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000571605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000573845.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000573845.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000573845.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000575302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000575302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000575302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000575302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000576408.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000576408.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000576408.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000576408.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000576408.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000576915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000576915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000576915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000576915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000576915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000576915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000576915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000576915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000576915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000576915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000576915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
ENST00000576915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
NM_001907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
NM_001907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
NM_001907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
NM_001907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
NM_001907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
NM_001907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
NM_001907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
NM_001907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
NM_001907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
NM_001907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
NM_001907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
NM_001907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011552.hg.1 |
NM_001907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16016684.hg.1 |
TC16002087.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16016684.hg.1 |
ENST00000319955.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16016684.hg.1 |
NM_001907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600193987.hg.1 |
ENST00000571044.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600193987.hg.1 |
ENST00000571605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600193987.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600193987.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600193987.hg.1 |
ENST00000576915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600193987.hg.1 |
NM_001907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600193987.hg.1 |
ENST00000571044.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600193987.hg.1 |
ENST00000571605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600193987.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600193987.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600193987.hg.1 |
ENST00000576915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600193987.hg.1 |
NM_001907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600193987.hg.1 |
TC1600011552.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000571044.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000571044.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000571044.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000571044.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000571044.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000571044.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000571044.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000571044.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000571605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000571605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000571605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000571605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000571605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000571605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000571605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000571605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000572144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000573845.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000573845.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000573845.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000574481.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000575302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000575302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000575302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000575302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000576408.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000576408.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000576408.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000576408.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000576408.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000576915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000576915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000576915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000576915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000576915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000576915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000576915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000576915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000576915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000576915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000576915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
ENST00000576915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
NM_001907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
NM_001907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
NM_001907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
NM_001907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
NM_001907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
NM_001907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
NM_001907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
NM_001907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
NM_001907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
NM_001907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
NM_001907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
NM_001907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011552.hg.1 |
NM_001907.2 |
Refseq |