Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027145.hg.1 |
TC06002152.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027145.hg.1 |
ENST00000367735.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027145.hg.1 |
ENST00000367739.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027145.hg.1 |
ENST00000418947.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027145.hg.1 |
ENST00000543628.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027145.hg.1 |
NM_000416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000367739.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000367739.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000367739.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000367739.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000367739.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000367739.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000367739.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000367739.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000367739.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000367739.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000367739.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000367739.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000367739.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000367739.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000414770.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000414770.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000414770.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000414770.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000414770.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000414770.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000414770.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000414770.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000414770.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000458076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000458076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000458076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000458076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000458076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000458076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000458076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000458076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000458076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000458076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000478333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000478333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000478333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000478333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000478333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000543628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000543628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000543628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000543628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000543628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000543628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000543628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000543628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000543628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000543628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000543628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000543628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
ENST00000543628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
NM_000416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
NM_000416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
NM_000416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
NM_000416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
NM_000416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
NM_000416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
NM_000416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
NM_000416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
NM_000416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
NM_000416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
NM_000416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
NM_000416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
NM_000416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013300.hg.1 |
NM_000416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000367739.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000367739.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000367739.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000367739.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000367739.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000367739.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000367739.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000367739.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000367739.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000367739.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000367739.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000367739.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000367739.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000367739.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000414770.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000414770.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000414770.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000414770.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000414770.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000414770.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000414770.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000414770.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000414770.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000458076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000458076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000458076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000458076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000458076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000458076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000458076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000458076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000458076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000458076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000478333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000478333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000478333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000478333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000478333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000543628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000543628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000543628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000543628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000543628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000543628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000543628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000543628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000543628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000543628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000543628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000543628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
ENST00000543628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
NM_000416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
NM_000416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
NM_000416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
NM_000416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
NM_000416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
NM_000416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
NM_000416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
NM_000416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
NM_000416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
NM_000416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
NM_000416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
NM_000416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
NM_000416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013300.hg.1 |
NM_000416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600192058.hg.1 |
ENST00000367739.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600192058.hg.1 |
ENST00000414770.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600192058.hg.1 |
ENST00000543628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600192058.hg.1 |
NM_000416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600192058.hg.1 |
ENST00000367739.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600192058.hg.1 |
ENST00000414770.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600192058.hg.1 |
ENST00000543628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600192058.hg.1 |
NM_000416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600192058.hg.1 |
TC0600013300.hg.1 |
Clariom D TC |