Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011005.hg.1 |
TC06000947.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011005.hg.1 |
ENST00000368455.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011005.hg.1 |
ENST00000452194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011005.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011005.hg.1 |
NM_001243094.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011005.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000465214.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000465214.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000465214.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000465214.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
ENST00000465214.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_001243094.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_001243094.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_001243094.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_001243094.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_001243094.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_001243094.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_001243094.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_001243094.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_001243094.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_001243094.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_001243094.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000465214.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000465214.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000465214.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000465214.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
ENST00000465214.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_001243094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_001243094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_001243094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_001243094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_001243094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_001243094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_001243094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_001243094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_001243094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_001243094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_001243094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009322.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163831.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163831.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163831.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163831.hg.1 |
NM_001243094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163831.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163831.hg.1 |
ENST00000368455.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163831.hg.1 |
ENST00000452194.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163831.hg.1 |
NM_001135564.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163831.hg.1 |
NM_001243094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163831.hg.1 |
NM_004506.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163831.hg.1 |
TC0600009322.hg.1 |
Clariom D TC |