Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000884.hg.1 |
TC21001059.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000884.hg.1 |
ENST00000290200.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000884.hg.1 |
ENST00000539894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000884.hg.1 |
NM_000628.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000290200.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000290200.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000290200.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000290200.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000290200.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000290200.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000290200.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000290200.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000290200.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000290200.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000290200.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000422891.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000422891.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000422891.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000422891.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000422891.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000422891.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000422891.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000451065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000451065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000451065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000451065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000451065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000451065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000451065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000493295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000493295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000493295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000493295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000493295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000493295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000493295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000493295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000493295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000493295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000498371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000498371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000498371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
ENST00000498371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
NM_000628.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
NM_000628.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
NM_000628.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
NM_000628.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
NM_000628.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
NM_000628.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
NM_000628.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
NM_000628.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
NM_000628.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
NM_000628.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008495.hg.1 |
NM_000628.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000290200.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000290200.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000290200.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000290200.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000290200.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000290200.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000290200.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000290200.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000290200.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000290200.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000290200.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000422891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000422891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000422891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000422891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000422891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000422891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000422891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000451065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000451065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000451065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000451065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000451065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000451065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000451065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000493295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000493295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000493295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000493295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000493295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000493295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000493295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000493295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000493295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000493295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000498371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000498371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000498371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
ENST00000498371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
NM_000628.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
NM_000628.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
NM_000628.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
NM_000628.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
NM_000628.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
NM_000628.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
NM_000628.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
NM_000628.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
NM_000628.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
NM_000628.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008495.hg.1 |
NM_000628.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100157920.hg.1 |
TC2100008495.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100157920.hg.1 |
ENST00000290200.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157920.hg.1 |
ENST00000422891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157920.hg.1 |
ENST00000451065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157920.hg.1 |
ENST00000493295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157920.hg.1 |
ENST00000498371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157920.hg.1 |
NM_000628.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100157920.hg.1 |
ENST00000290200.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157920.hg.1 |
ENST00000422891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157920.hg.1 |
ENST00000451065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157920.hg.1 |
ENST00000493295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157920.hg.1 |
ENST00000498371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157920.hg.1 |
NM_000628.4 |
Refseq |