Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02041560.hg.1 |
TC02002591.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02041560.hg.1 |
ENST00000295113.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02041560.hg.1 |
NM_001463.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015136.hg.1 |
ENST00000295113.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015136.hg.1 |
ENST00000295113.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015136.hg.1 |
ENST00000295113.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015136.hg.1 |
ENST00000295113.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015136.hg.1 |
ENST00000295113.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015136.hg.1 |
ENST00000295113.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015136.hg.1 |
ENST00000295113.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015136.hg.1 |
ENST00000295113.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015136.hg.1 |
ENST00000295113.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015136.hg.1 |
ENST00000295113.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015136.hg.1 |
ENST00000295113.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015136.hg.1 |
NM_001463.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015136.hg.1 |
NM_001463.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015136.hg.1 |
NM_001463.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015136.hg.1 |
NM_001463.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015136.hg.1 |
NM_001463.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015136.hg.1 |
NM_001463.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015136.hg.1 |
NM_001463.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015136.hg.1 |
NM_001463.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015136.hg.1 |
NM_001463.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015136.hg.1 |
NM_001463.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200210916.hg.1 |
ENST00000295113.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200210916.hg.1 |
NM_001463.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200210916.hg.1 |
ENST00000295113.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200210916.hg.1 |
NM_001463.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200210916.hg.1 |
TC0200015136.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200015136.hg.1 |
ENST00000295113.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015136.hg.1 |
ENST00000295113.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015136.hg.1 |
ENST00000295113.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015136.hg.1 |
ENST00000295113.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015136.hg.1 |
ENST00000295113.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015136.hg.1 |
ENST00000295113.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015136.hg.1 |
ENST00000295113.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015136.hg.1 |
ENST00000295113.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015136.hg.1 |
ENST00000295113.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015136.hg.1 |
ENST00000295113.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015136.hg.1 |
ENST00000295113.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015136.hg.1 |
NM_001463.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015136.hg.1 |
NM_001463.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015136.hg.1 |
NM_001463.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015136.hg.1 |
NM_001463.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015136.hg.1 |
NM_001463.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015136.hg.1 |
NM_001463.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015136.hg.1 |
NM_001463.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015136.hg.1 |
NM_001463.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015136.hg.1 |
NM_001463.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015136.hg.1 |
NM_001463.3 |
Refseq |