Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013151.hg.1 |
TC01000826.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013151.hg.1 |
ENST00000212369.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013151.hg.1 |
ENST00000370558.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013151.hg.1 |
ENST00000482504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013151.hg.1 |
ENST00000535010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013151.hg.1 |
NM_001206651.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013151.hg.1 |
NM_001206652.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013151.hg.1 |
NM_001206653.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013151.hg.1 |
NM_016009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163719.hg.1 |
ENST00000370558.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163719.hg.1 |
ENST00000482504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163719.hg.1 |
ENST00000535010.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163719.hg.1 |
ENST00000602603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163719.hg.1 |
ENST00000616170.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163719.hg.1 |
NM_001206651.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163719.hg.1 |
NM_001206652.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163719.hg.1 |
NM_001206653.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163719.hg.1 |
NM_016009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163719.hg.1 |
ENST00000370558.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163719.hg.1 |
ENST00000482504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163719.hg.1 |
ENST00000535010.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163719.hg.1 |
ENST00000602603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163719.hg.1 |
ENST00000616170.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163719.hg.1 |
NM_001206651.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163719.hg.1 |
NM_001206652.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163719.hg.1 |
NM_001206653.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163719.hg.1 |
NM_016009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163719.hg.1 |
TC0100008927.hg.1 |
Clariom D TC |