Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
ENST00000392039.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
ENST00000392039.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
ENST00000392039.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
ENST00000392039.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
ENST00000392039.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
ENST00000392040.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
ENST00000392040.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
ENST00000392040.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
ENST00000392040.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
ENST00000392040.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
ENST00000392040.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
ENST00000392040.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
ENST00000392040.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
ENST00000392040.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
ENST00000438398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
ENST00000438398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
ENST00000438398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
ENST00000444078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
ENST00000444078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
ENST00000444078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
ENST00000444078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
ENST00000444078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
ENST00000444078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
ENST00000444078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
ENST00000444078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
ENST00000622008.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
ENST00000622008.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
ENST00000622008.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
ENST00000622008.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
ENST00000622008.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
ENST00000622008.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
ENST00000622008.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
ENST00000622008.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
NM_005683.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
NM_005683.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
NM_005683.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
NM_005683.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
NM_005683.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
NM_005683.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
NM_005683.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
NM_005683.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016000.hg.1 |
NM_005683.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
ENST00000392039.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
ENST00000392039.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
ENST00000392039.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
ENST00000392039.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
ENST00000392039.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
ENST00000392040.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
ENST00000392040.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
ENST00000392040.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
ENST00000392040.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
ENST00000392040.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
ENST00000392040.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
ENST00000392040.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
ENST00000392040.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
ENST00000392040.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
ENST00000438398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
ENST00000438398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
ENST00000438398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
ENST00000444078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
ENST00000444078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
ENST00000444078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
ENST00000444078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
ENST00000444078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
ENST00000444078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
ENST00000444078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
ENST00000444078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
ENST00000622008.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
ENST00000622008.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
ENST00000622008.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
ENST00000622008.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
ENST00000622008.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
ENST00000622008.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
ENST00000622008.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
ENST00000622008.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
NM_005683.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
NM_005683.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
NM_005683.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
NM_005683.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
NM_005683.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
NM_005683.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
NM_005683.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
NM_005683.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016000.hg.1 |
NM_005683.3 |
Refseq |