Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017767.hg.1 |
ENST00000269878.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017767.hg.1 |
NM_054113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017767.hg.1 |
TC19001264.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017769.hg.1 |
ENST00000269878.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017769.hg.1 |
ENST00000541493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017769.hg.1 |
NM_054113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017769.hg.1 |
TC19001264.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
ENST00000269878.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
ENST00000269878.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
ENST00000269878.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
ENST00000269878.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
ENST00000269878.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
ENST00000269878.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
ENST00000269878.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
ENST00000269878.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
ENST00000379859.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
ENST00000379859.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
ENST00000379859.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
ENST00000379859.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
ENST00000541493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
ENST00000541493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
ENST00000541493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
ENST00000541493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
ENST00000597251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
ENST00000597251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
ENST00000597251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
ENST00000597251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
ENST00000597251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
ENST00000597251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
NM_001300922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
NM_001300922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
NM_001300922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
NM_001300922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
NM_001300922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
NM_001300922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
NM_054113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
NM_054113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
NM_054113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
NM_054113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
NM_054113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
NM_054113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
NM_054113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009906.hg.1 |
NM_054113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017770.hg.1 |
ENST00000269878.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017770.hg.1 |
ENST00000379859.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017770.hg.1 |
ENST00000541493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017770.hg.1 |
NM_054113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017770.hg.1 |
TC19001264.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
ENST00000269878.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
ENST00000269878.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
ENST00000269878.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
ENST00000269878.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
ENST00000269878.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
ENST00000269878.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
ENST00000269878.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
ENST00000269878.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
ENST00000379859.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
ENST00000379859.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
ENST00000379859.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
ENST00000379859.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
ENST00000541493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
ENST00000541493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
ENST00000541493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
ENST00000541493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
ENST00000597251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
ENST00000597251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
ENST00000597251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
ENST00000597251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
ENST00000597251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
ENST00000597251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
NM_001300922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
NM_001300922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
NM_001300922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
NM_001300922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
NM_001300922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
NM_001300922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
NM_054113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
NM_054113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
NM_054113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
NM_054113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
NM_054113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
NM_054113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
NM_054113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009906.hg.1 |
NM_054113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900181236.hg.1 |
ENST00000269878.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181236.hg.1 |
ENST00000541493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181236.hg.1 |
ENST00000597251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181236.hg.1 |
NM_054113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900181236.hg.1 |
TC1900009906.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900181236.hg.1 |
ENST00000269878.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181236.hg.1 |
ENST00000541493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181236.hg.1 |
ENST00000597251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181236.hg.1 |
NM_054113.3 |
Refseq |