Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001184.3 | NP_001175.2 | 20 - 21 | 3941 | 144 | |
XM_011512924.1 | XP_011511226.1 | 20 - 21 | 3908 | 144 | ||
XM_011512925.1 | XP_011511227.1 | 19 - 20 | 3716 | 144 | ||
XM_011512926.2 | XP_011511228.1 | 20 - 21 | 3908 | 144 | ||
XM_017006643.1 | XP_016862132.1 | 19 - 20 | 3716 | 144 | ||
GenBank mRNA | U49844.1 | - | 20 - 21 | 3924 | 144 | |
U76308.1 | - | 20 - 21 | 3898 | 144 | ||
Y09077.1 | - | 20 - 21 | 3898 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Inventoried Probe spans exons |