Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010464.hg.1 |
ENST00000369108.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010464.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010464.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010464.hg.1 |
TC10000865.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
ENST00000369108.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
ENST00000369108.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
ENST00000369108.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
ENST00000369108.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
ENST00000369108.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
ENST00000369108.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
ENST00000369108.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
ENST00000369108.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
ENST00000369108.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
ENST00000369108.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
ENST00000369108.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
ENST00000369108.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
ENST00000369108.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
ENST00000369108.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
ENST00000369108.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
ENST00000369108.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009067.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162099.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162099.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162099.hg.1 |
ENST00000392870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162099.hg.1 |
NM_005308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162099.hg.1 |
TC1000009067.hg.1 |
Clariom D TC |