Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024615.hg.1 |
TC04001807.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024615.hg.1 |
ENST00000308394.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024615.hg.1 |
ENST00000393585.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024615.hg.1 |
ENST00000393588.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024615.hg.1 |
ENST00000438467.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024615.hg.1 |
ENST00000517513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024615.hg.1 |
ENST00000523916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024615.hg.1 |
NM_004346.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024615.hg.1 |
NM_032991.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400186344.hg.1 |
ENST00000308394.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186344.hg.1 |
ENST00000393585.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186344.hg.1 |
ENST00000393588.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186344.hg.1 |
ENST00000447121.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186344.hg.1 |
ENST00000517513.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186344.hg.1 |
ENST00000523916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186344.hg.1 |
ENST00000613118.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186344.hg.1 |
NM_004346.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400186344.hg.1 |
NM_032991.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400186344.hg.1 |
ENST00000308394.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186344.hg.1 |
ENST00000393585.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186344.hg.1 |
ENST00000393588.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186344.hg.1 |
ENST00000447121.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186344.hg.1 |
ENST00000517513.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186344.hg.1 |
ENST00000523916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186344.hg.1 |
ENST00000613118.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186344.hg.1 |
NM_004346.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400186344.hg.1 |
NM_032991.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400186344.hg.1 |
TC0400012618.hg.1 |
Clariom D TC |