Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | PRH1-PRR4 readthrough | PRH1-PRR4 | PRH1 | Anti-sense | Chr. 12 - 10845849 - 11171625 on NCBI Build GRCh38 | |
Downstream | taste 2 receptor member 30 | TAS2R30 |
T2R30, T2R47, TAS2R47 |
36.4kb | Anti-sense | Chr. 12 - 11133285 - 11134244 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |