Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
ENST00000380031.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
ENST00000380031.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
ENST00000380031.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
ENST00000380031.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
ENST00000380031.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
ENST00000380031.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
ENST00000380031.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
ENST00000380032.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
ENST00000380032.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
ENST00000380032.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
ENST00000380032.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
ENST00000380032.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
ENST00000380032.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
ENST00000380032.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
ENST00000380032.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
ENST00000380032.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
ENST00000380032.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
ENST00000380032.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
ENST00000380032.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
ENST00000484994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
ENST00000484994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
ENST00000484994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
ENST00000484994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
ENST00000537675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
ENST00000537675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
ENST00000537675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
ENST00000537675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
ENST00000537675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
ENST00000537675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
ENST00000537675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
ENST00000537675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
ENST00000537675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
NM_001161585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
NM_001161585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
NM_001161585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
NM_001161585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
NM_001161585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
NM_001161585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
NM_001161585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
NM_001161585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
NM_001161585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
NM_001161585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
NM_001161585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
NM_020641.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
NM_020641.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
NM_020641.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
NM_020641.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
NM_020641.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
NM_020641.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
NM_020641.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
NM_020641.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
NM_020641.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
NM_020641.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
NM_020641.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009775.hg.1 |
NM_020641.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012973.hg.1 |
TC09000987.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012973.hg.1 |
ENST00000380032.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012973.hg.1 |
ENST00000537675.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012973.hg.1 |
NM_001161585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012973.hg.1 |
NM_020641.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166891.hg.1 |
ENST00000380032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166891.hg.1 |
ENST00000537675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166891.hg.1 |
NM_001161585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166891.hg.1 |
NM_020641.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166891.hg.1 |
ENST00000380032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166891.hg.1 |
ENST00000537675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166891.hg.1 |
NM_001161585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166891.hg.1 |
NM_020641.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166891.hg.1 |
TC0900009775.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
ENST00000380031.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
ENST00000380031.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
ENST00000380031.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
ENST00000380031.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
ENST00000380031.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
ENST00000380031.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
ENST00000380031.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
ENST00000380032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
ENST00000380032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
ENST00000380032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
ENST00000380032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
ENST00000380032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
ENST00000380032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
ENST00000380032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
ENST00000380032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
ENST00000380032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
ENST00000380032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
ENST00000380032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
ENST00000380032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
ENST00000484994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
ENST00000484994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
ENST00000484994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
ENST00000484994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
ENST00000537675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
ENST00000537675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
ENST00000537675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
ENST00000537675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
ENST00000537675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
ENST00000537675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
ENST00000537675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
ENST00000537675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
ENST00000537675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
NM_001161585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
NM_001161585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
NM_001161585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
NM_001161585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
NM_001161585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
NM_001161585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
NM_001161585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
NM_001161585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
NM_001161585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
NM_001161585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
NM_001161585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
NM_020641.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
NM_020641.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
NM_020641.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
NM_020641.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
NM_020641.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
NM_020641.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
NM_020641.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
NM_020641.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
NM_020641.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
NM_020641.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
NM_020641.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009775.hg.1 |
NM_020641.2 |
Refseq |