Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021533.hg.1 |
TC04001516.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021533.hg.1 |
ENST00000274026.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021533.hg.1 |
NM_001237.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
ENST00000274026.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
ENST00000274026.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
ENST00000274026.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
ENST00000274026.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
ENST00000274026.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
ENST00000274026.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
ENST00000274026.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
ENST00000274026.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
ENST00000274026.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
ENST00000274026.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
ENST00000274026.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
ENST00000274026.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
ENST00000618014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
ENST00000618014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
ENST00000618014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
ENST00000618014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
ENST00000618014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
ENST00000618014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
ENST00000618014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
ENST00000618014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
ENST00000618014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
ENST00000618014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
ENST00000618014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
ENST00000618014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
NM_001237.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
NM_001237.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
NM_001237.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
NM_001237.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
NM_001237.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
NM_001237.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
NM_001237.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
NM_001237.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
NM_001237.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
NM_001237.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
NM_001237.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011748.hg.1 |
NM_001237.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180822.hg.1 |
ENST00000274026.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180822.hg.1 |
ENST00000618014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180822.hg.1 |
NM_001237.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180822.hg.1 |
ENST00000274026.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180822.hg.1 |
ENST00000618014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180822.hg.1 |
NM_001237.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180822.hg.1 |
TC0400011748.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
ENST00000274026.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
ENST00000274026.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
ENST00000274026.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
ENST00000274026.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
ENST00000274026.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
ENST00000274026.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
ENST00000274026.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
ENST00000274026.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
ENST00000274026.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
ENST00000274026.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
ENST00000274026.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
ENST00000274026.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
ENST00000618014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
ENST00000618014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
ENST00000618014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
ENST00000618014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
ENST00000618014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
ENST00000618014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
ENST00000618014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
ENST00000618014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
ENST00000618014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
ENST00000618014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
ENST00000618014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
ENST00000618014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
NM_001237.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
NM_001237.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
NM_001237.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
NM_001237.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
NM_001237.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
NM_001237.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
NM_001237.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
NM_001237.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
NM_001237.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
NM_001237.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
NM_001237.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011748.hg.1 |
NM_001237.4 |
Refseq |