Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015824.hg.1 |
TC0X001174.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015824.hg.1 |
ENST00000358075.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015824.hg.1 |
ENST00000373336.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015824.hg.1 |
NM_032121.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000358075.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000358075.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000358075.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000358075.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000358075.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000358075.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000358075.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000358075.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000358075.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000358075.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000358075.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000358075.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000358075.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000358075.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000373336.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000373336.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000373336.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000373336.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000476168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000476168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000476168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000476168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000476168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000476168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000610432.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000610432.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000610432.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000610432.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000610432.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000610432.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000610432.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000610432.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000610432.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000610432.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000610432.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000610432.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000610432.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000618282.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000618282.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000618282.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000618282.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000618282.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000618282.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000618282.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000618282.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000618282.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
ENST00000618282.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
NM_032121.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
NM_032121.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
NM_032121.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
NM_032121.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
NM_032121.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
NM_032121.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
NM_032121.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
NM_032121.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
NM_032121.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
NM_032121.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
NM_032121.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
NM_032121.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010130.hg.1 |
NM_032121.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015823.hg.1 |
TC0X001174.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015823.hg.1 |
ENST00000358075.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015823.hg.1 |
ENST00000373336.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015823.hg.1 |
ENST00000453109.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015823.hg.1 |
NM_032121.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169178.hg.1 |
ENST00000358075.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169178.hg.1 |
ENST00000373336.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169178.hg.1 |
ENST00000476168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169178.hg.1 |
ENST00000610432.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169178.hg.1 |
ENST00000618282.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169178.hg.1 |
NM_032121.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169178.hg.1 |
ENST00000358075.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169178.hg.1 |
ENST00000373336.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169178.hg.1 |
ENST00000476168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169178.hg.1 |
ENST00000610432.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169178.hg.1 |
ENST00000618282.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169178.hg.1 |
NM_032121.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169178.hg.1 |
TC0X00010130.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000358075.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000358075.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000358075.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000358075.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000358075.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000358075.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000358075.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000358075.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000358075.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000358075.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000358075.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000358075.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000358075.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000358075.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000373336.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000373336.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000373336.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000373336.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000476168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000476168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000476168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000476168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000476168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000476168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000610432.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000610432.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000610432.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000610432.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000610432.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000610432.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000610432.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000610432.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000610432.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000610432.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000610432.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000610432.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000610432.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000618282.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000618282.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000618282.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000618282.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000618282.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000618282.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000618282.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000618282.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000618282.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
ENST00000618282.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
NM_032121.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
NM_032121.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
NM_032121.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
NM_032121.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
NM_032121.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
NM_032121.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
NM_032121.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
NM_032121.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
NM_032121.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
NM_032121.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
NM_032121.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
NM_032121.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010130.hg.1 |
NM_032121.5 |
Refseq |