Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000538042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000538042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000538042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000538042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000538595.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000538595.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000538595.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000538595.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000538595.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000538595.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000538595.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000538595.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000541041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000541041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000541041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000541041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000542105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000542105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000542105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000542105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000542105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000542359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000542359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000542359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000542359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000542359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000542359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000543422.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000543422.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000543422.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000543422.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000543422.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000545464.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000545464.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000545464.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000545464.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000545464.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000545464.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000545464.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000545464.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000545464.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
ENST00000545464.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11022386.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11022386.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11022386.hg.1 |
TC11001898.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100184950.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100184950.hg.1 |
ENST00000543422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100184950.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100184950.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100184950.hg.1 |
ENST00000543422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100184950.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100184950.hg.1 |
TC1100011144.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000255681.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000538042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000538042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000538042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000538042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000538595.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000538595.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000538595.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000538595.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000538595.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000538595.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000538595.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000538595.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000541041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000541041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000541041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000541041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000542105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000542105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000542105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000542105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000542105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000542359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000542359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000542359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000542359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000542359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000542359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000543422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000543422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000543422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000543422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000543422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000545464.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000545464.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000545464.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000545464.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000545464.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000545464.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000545464.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000545464.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000545464.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
ENST00000545464.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011144.hg.1 |
NM_014067.3 |
Refseq |