Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008485.hg.1 |
TC03000394.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008485.hg.1 |
ENST00000295900.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008485.hg.1 |
ENST00000398590.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008485.hg.1 |
ENST00000484332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008485.hg.1 |
ENST00000487717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008485.hg.1 |
ENST00000488239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008485.hg.1 |
ENST00000538065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008485.hg.1 |
ENST00000539129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008485.hg.1 |
NM_000333.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008485.hg.1 |
NM_001128149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008485.hg.1 |
NM_001177387.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155511.hg.1 |
ENST00000295900.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155511.hg.1 |
ENST00000474112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155511.hg.1 |
ENST00000475897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155511.hg.1 |
ENST00000484332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155511.hg.1 |
ENST00000487717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155511.hg.1 |
ENST00000488239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155511.hg.1 |
ENST00000538065.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155511.hg.1 |
NM_000333.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155511.hg.1 |
NM_001128149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155511.hg.1 |
NM_001177387.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155511.hg.1 |
ENST00000295900.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155511.hg.1 |
ENST00000474112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155511.hg.1 |
ENST00000475897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155511.hg.1 |
ENST00000484332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155511.hg.1 |
ENST00000487717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155511.hg.1 |
ENST00000488239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155511.hg.1 |
ENST00000538065.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155511.hg.1 |
NM_000333.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155511.hg.1 |
NM_001128149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155511.hg.1 |
NM_001177387.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155511.hg.1 |
TC0300007679.hg.1 |
Clariom D TC |