Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000310054.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000310054.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000310054.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000310054.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000310054.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000310054.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000310054.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000310054.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000310054.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000310054.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000310054.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000310054.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000310054.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000310054.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000310054.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000310054.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000593545.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000593545.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000593545.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000593545.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000593545.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000593545.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000593545.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000593545.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000593545.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000593545.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000593890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000593890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000593890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000593890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000593890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000595590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000595590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000595590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000595590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000595590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000595590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000595590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000597754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000597754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000597754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000597754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000597754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000600561.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000600561.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000600561.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
ENST00000600561.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
NM_030622.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
NM_030622.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
NM_030622.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
NM_030622.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
NM_030622.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
NM_030622.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
NM_030622.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
NM_030622.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
NM_030622.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
NM_030622.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
NM_030622.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
NM_030622.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
NM_030622.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
NM_030622.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008141.hg.1 |
NM_030622.7 |
Refseq |