Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000261206.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000261206.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000261206.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000261206.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000261206.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000261206.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000261206.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000261206.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000261206.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000261206.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000261206.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000261206.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000261206.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000261206.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000261206.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000261206.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000261206.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000261206.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000261206.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000261206.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000546664.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000546664.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000546664.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000546664.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548058.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548058.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548058.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548058.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548058.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548058.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548058.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548058.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548058.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548058.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548058.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548058.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548058.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548058.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548058.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548058.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548058.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548058.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548058.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548058.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548058.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548058.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548058.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548058.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000548387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000551745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000551745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000551745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000551745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000551745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000551745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
ENST00000551745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
NM_024560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
NM_024560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
NM_024560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
NM_024560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
NM_024560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
NM_024560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
NM_024560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
NM_024560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
NM_024560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
NM_024560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
NM_024560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
NM_024560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
NM_024560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
NM_024560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
NM_024560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
NM_024560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
NM_024560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
NM_024560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
NM_024560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
NM_024560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
NM_024560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012680.hg.1 |
NM_024560.3 |
Refseq |