Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011658.hg.1 |
ENST00000370110.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011658.hg.1 |
ENST00000370110.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011658.hg.1 |
ENST00000370110.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011658.hg.1 |
ENST00000370110.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011658.hg.1 |
ENST00000370110.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011658.hg.1 |
ENST00000370110.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011658.hg.1 |
ENST00000370110.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011658.hg.1 |
ENST00000462391.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011658.hg.1 |
ENST00000462391.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011658.hg.1 |
ENST00000462391.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011658.hg.1 |
ENST00000462391.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011658.hg.1 |
ENST00000462391.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011658.hg.1 |
ENST00000468544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011658.hg.1 |
ENST00000468544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011658.hg.1 |
ENST00000468544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011658.hg.1 |
ENST00000468544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011658.hg.1 |
ENST00000474993.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011658.hg.1 |
ENST00000474993.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011658.hg.1 |
ENST00000474993.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011658.hg.1 |
ENST00000474993.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011658.hg.1 |
ENST00000474993.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011658.hg.1 |
ENST00000474993.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011658.hg.1 |
NM_006993.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011658.hg.1 |
NM_006993.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011658.hg.1 |
NM_006993.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011658.hg.1 |
NM_006993.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011658.hg.1 |
NM_006993.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011658.hg.1 |
NM_006993.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011658.hg.1 |
NM_006993.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020900.hg.1 |
ENST00000370110.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020900.hg.1 |
ENST00000474993.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020900.hg.1 |
NM_006993.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020900.hg.1 |
TC10001612.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1000011658.hg.1 |
ENST00000370110.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011658.hg.1 |
ENST00000370110.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011658.hg.1 |
ENST00000370110.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011658.hg.1 |
ENST00000370110.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011658.hg.1 |
ENST00000370110.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011658.hg.1 |
ENST00000370110.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011658.hg.1 |
ENST00000370110.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011658.hg.1 |
ENST00000462391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011658.hg.1 |
ENST00000462391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011658.hg.1 |
ENST00000462391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011658.hg.1 |
ENST00000462391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011658.hg.1 |
ENST00000462391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011658.hg.1 |
ENST00000468544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011658.hg.1 |
ENST00000468544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011658.hg.1 |
ENST00000468544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011658.hg.1 |
ENST00000468544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011658.hg.1 |
ENST00000474993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011658.hg.1 |
ENST00000474993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011658.hg.1 |
ENST00000474993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011658.hg.1 |
ENST00000474993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011658.hg.1 |
ENST00000474993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011658.hg.1 |
ENST00000474993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011658.hg.1 |
NM_006993.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011658.hg.1 |
NM_006993.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011658.hg.1 |
NM_006993.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011658.hg.1 |
NM_006993.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011658.hg.1 |
NM_006993.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011658.hg.1 |
NM_006993.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011658.hg.1 |
NM_006993.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179722.hg.1 |
ENST00000370110.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179722.hg.1 |
ENST00000462391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179722.hg.1 |
ENST00000468544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179722.hg.1 |
ENST00000474993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179722.hg.1 |
NM_006993.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179722.hg.1 |
ENST00000370110.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179722.hg.1 |
ENST00000462391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179722.hg.1 |
ENST00000468544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179722.hg.1 |
ENST00000474993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179722.hg.1 |
NM_006993.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179722.hg.1 |
TC1000011658.hg.1 |
Clariom D TC |